Skip to Content
MilliporeSigma
All Photos(15)

Documents

ZRB05636

Sigma-Aldrich

Anti-phospho-Histone H2A.X (Ser139) Antibody, clone 6L16, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

H2a/x, Histone H2A.X

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

6L16, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 15.15 kDa
observed mol wt ~15 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable using 1:100
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable using 1:100
inhibition assay: suitable using 1:1,000 (peptide)
western blot: suitable using 1:1,000

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

phosphorylation (pSer139)

Gene Information

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 6L16 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Histone H2A.X phosphorylated on serine 139.

Immunogen

KLH-conjugated linear peptide corresponding to 9 amino acids surrounding phospho-serine 139 of human Histone H2A.X.

Application

Anti-phospho-Histone H2A.X (Ser139), clone 6L16 ZooMAb, is a recombinant Rabbit Monoclonal Antibody that specifically targets phospho Histone H2AX (ser139) and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), Peptide Inhibition Assay, and Western Blotting.
Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected phospho-Histone H2A.X (Ser139) in human testis tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected phospho-Histone H2A.X (Ser139) in HeLa treated with Staurosporin.

Peptide Inhibition Analysis: A 1:1,000 dilution from a representative lot was used with Jurkat cell lysates treated with staurosporin for peptide block analysis.

Target description

Histone H2AX (UniProt: P16104; also known as H2a/x, Histone H2A.X) is encoded by the H2AFX (also known as H2AX) gene (Gene ID: 3014) in human. Histones are highly conserved basic nuclear proteins that are responsible for the nucleosome structure of chromatin in eukaryotes. They play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which DNA is wrapped in repeating units, called nucleosomes, which limits DNA accessibility to the cellular machineries that require DNA as a template. The histone H2A.X is a variant member of the H2A family of histones that is distinguished from other H2A histones by a unique carboxy-terminal sequence. This unique sequence is highly conserved throughout eukaryotic evolution and is rapidly phosphorylated by ATM or ATR at Serine 139 in mammals in response to DNA double-strand breaks. H2A.X phosphorylation is important in the formation of a stable repair complex at the site of DNA damage. Phosphorylation of H2A.X is important in the formation of a stable repair complex at the site of DNA damage. This phosphorylation is a very rapid response to DNA damage, occurring within as little as one minute after exposure to ionizing radiation. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

30 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian Gao et al.
Cell death & disease, 10(12), 957-957 (2019-12-22)
Radiation-induced lung injury (RILI) is one of the most common and fatal complications of thoracic radiotherapy, whereas no effective interventions are available. Andrographolide, an active component extracted from Andrographis paniculate, is prescribed as a treatment for upper respiratory tract infection.
Ria Winkelmann et al.
BMC cancer, 22(1), 1352-1352 (2022-12-24)
Penile cancer is a rare disease in demand for new therapeutic options. Frequently used combination chemotherapy with 5 fluorouracil (5-FU) and cisplatin (CDDP) in patients with metastatic penile cancer mostly results in the development of acquired drug resistance. Availability of
Blanca Ocampo-García et al.
International journal of molecular sciences, 24(23) (2023-12-09)
Prostate-specific membrane antigens (PSMAs) are frequently overexpressed in both tumor stromal endothelial cells and malignant cells (stromal/tumor cells) of various cancers. The RGD (Arg-Gly-Asp) peptide sequence can specifically detect integrins involved in tumor angiogenesis. This study aimed to preclinically evaluate
Yu-Chi Chen et al.
Molecules (Basel, Switzerland), 25(17) (2020-09-03)
A clerodane diterpene compound 16-hydroxycleroda-3,13-dien-15,16-olide (CD) is considered a therapeutic agent with pharmacological activities. The present study investigated the mechanisms of CD-induced apoptosis in T24 human bladder cancer cells. CD inhibited cell proliferation in a concentration and time-dependent manner. CD-induced

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Protocols

Follow this short antibody reconstitution protocol and see how easy it is to reconstitute ZooMAb® recombinant antibodies.

Follow this short antibody reconstitution protocol and see how easy it is to reconstitute ZooMAb® recombinant antibodies.

Follow this short antibody reconstitution protocol and see how easy it is to reconstitute ZooMAb® recombinant antibodies.

Follow this short antibody reconstitution protocol and see how easy it is to reconstitute ZooMAb® recombinant antibodies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service