Skip to Content
MilliporeSigma
All Photos(3)

Documents

PLA0264

Sigma-Aldrich

Rabbit anti-Ku80 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

80kDa, 86 kDa subunit of Ku antigen, ATP-dependent DNA helicase 2 subunit 2, ATP-dependent DNA helicase II 80 kDa subunit, CTC box-binding factor 85 kDa subunit, CTC85, CTCBF, DNA repair protein XRCC5, KARP-1, KARP1, KU80, KUB2, Ku autoantigen, Ku86, NFIV, TLAA, X-ray repair complementing defective repair in Chinese hamster cells 5, double-strand-break rejoining, lupus Ku autoantigen protein p86, nuclear factor IV, thyroid-lupus autoantigen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human

technique(s)

immunohistochemistry: 1:100- 1:500
immunoprecipitation (IP): 2-5 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_066964.1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... Ku80(7520)

Immunogen

The epitope recognized by PLA0264 maps to a region between residue 150 and 200 of human Lupus Ku autoantigen protein p86 using the numbering given in entry NP_066964.1 (GeneID 7520).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

Ku80 is the 80 kDa subunit of the lupus Ku autoantigen. The lupus Ku autoantigen was identified in individuals with systemic lupus erythematosus (SLE) and related disorders. The Ku antigen is a heterodimer of Ku70 and Ku80. The Ku70/Ku80 complex functions as a single-stranded DNA-dependent ATP-dependent helicase. The complex may be involved in the repair of nonhomologous DNA ends such as that required for double-strand break repair, transposition, and V(D)J recombination [taken from the Universal Protein Resource (UniProt) www.uniprot.org/uniprot/P13010].

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dan Wang et al.
Cell death and differentiation, 27(12), 3337-3353 (2020-06-27)
Despite recent advances in our understanding of the function of long noncoding RNAs (lncRNAs), their roles and functions in DNA repair pathways remain poorly understood. By screening a panel of uncharacterized lncRNAs to identify those whose transcription is induced by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service