Skip to Content
MilliporeSigma
All Photos(9)

Key Documents

ZMS1066

Sigma-Aldrich

Anti-LINE-1 ORF1p Antibody, clone 4H1 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

4H1, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt ~40.06 kDa
observed mol wt ~40 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 4H1 is a ZooMAb® mouse recombinant monoclonal antibody that specifically detects LINE-1 retrotransposable element ORF1 protein. It targets an epitope within 10 amino acids from the N-terminal region.

Immunogen

A linear peptide corresponding to 10 amino acids from the N-terminal region of human LINE-1 ORF1 protein.

Application

Quality Control Testing

Evaluated by Western Blotting in MCF-7 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected LINE-1 ORF1p in MCF-7 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected LINE-1 ORF1p in NTERA-2 cell lysate.

Flow Cytometry Analysis: 0.1 μg from a representative lot detected LINE-1 ORF1p in one million MCF-7 cells.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected LINE-1 ORF1p in MCF-7 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-LINE-1 ORF1p, clone 4H1 ZooMAb®, Cat. No. ZMS1066, is a recombinant Mouse monoclonal antibody that specifically targets LINE-1 ORF1p and is tested for use in Flow Cytometry, Immunocytochemistry, and Western Blotting.

Target description

LINE-1 retrotransposable element ORF1 protein (UniProt: Q9UN81; also known L1ORF1p, LINE retrotransposable element 1, LINE1 retrotransposable element 1) is encoded by the LIRE1 (also known as LRE1) gene in human. LINE1-ORF 1P is a homotrimeric protein of the transposase 22 family that can also form larger homooligomers. It is mainly a cytoplasmic protein and is enriched in stress granules, but may also be detected in the nucleolus. It contains a coiled coil domain that mediates its homotrimerization. Its RNA recognition motif (RRM) (aa 157-252) in the C-terminal domain are required for its proper RNA-binding activity. LINE1-ORF 1P serves as a nucleic acid-binding protein that is essential for retrotransposition of LINE-1 elements in the genome. It is also reported to function as a nucleic acid chaperone binding its own transcript and therefore preferentially mobilizing the transcript from which they are encoded. Retrotransposition is a shown to be a complex process that involves transcription of the LINE1, transport of its RNA to the cytoplasm, translation of the bicistronic RNA, formation of a ribonucleoprotein particle, its re-import to the nucleus and then target-primed reverse transcription at the integration site. LINE1-ORF 1P expression is consistently detected in p53 mutant tumors, but, little or no expression is detected in Wilms tumors that were normal for p53. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Goodier, JL et al. (2013). Nucleic Acids Res. 41(15): 7401-7419).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service