Key Documents
Z765589
Tube rack for Benchmixer XL multi-tube vortexer
Holds 50 x 15 mL tubes
About This Item
UNSPSC Code:
41122804
NACRES:
NB.77
description
15 mL QuEChERS Extraction Tube Shaker Rack
feature
Holds 50 x 15 mL tubes
packaging
pkg of 1 ea
manufacturer/tradename
Benchmark Scientific BV1010-150
General description
Benchmixer XL vortexer head adapters to hold multiple tubes.
This rack enables the use of 50 x 15 mL tubes in the BenchMixer XL heavy-duty hands-free mixing vortex mixer. Ideal for the QuEChERS pesticide analysis method.
Legal Information
Benchmixer is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)