Z756466
PrepSafe™ microcentrifuge tube mini floating rack
yellow (neon)
Synonym(s):
rack test tube, test tube racks, tube racks
About This Item
Recommended Products
material
yellow (neon)
feature
Holds 12 x 1.5/2 mL tubes
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art F18742-0127
O.D.
95 mm (3.75 in.)
diam.
95 mm
Looking for similar products? Visit Product Comparison Guide
General description
- Holds up to twelve 1.5 mL or 2.0 mL microcentrifuge tubes in numbered locations
- Raised handle for easy lifting without agitating samples
- Stable feet keep racks upright on the bench
- Use in water baths, ice or on the benchtop
- 95 mm outer diameter fits most 1000 mL beakers
- Also available with vortex attachment and in 3 other colors
- Steam autoclavable at 121°C (250°F)
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service