Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z743673

Laser Tough-Tags microcentrifuge tube labels, kiss cut with square corners

white, pkg of 2520 labels 20 sheets of 126 labels, L × W 1.28 in. × 0.50 in.

Synonym(s):

solvent resistant labels, temperature resistant labels

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
55121611
NACRES:
NB.35

packaging

pkg of 2520 labels 20 sheets of 126 labels

manufacturer/tradename

Diversified Biotech TTLW-KK-20

L × W

1.28 in. × 0.50 in.

color

white

Looking for similar products? Visit Product Comparison Guide

General description

Laboratory labels formatted for laser printers. Tough-Tags are chemically inert laboratory labels that strongly adhere to microtubes, test tubes, storage boxes and microscope slides. They resist organic solvents and caustic agents making them perfect for accurately identifying important samples stored in moderate freezer temperatures.

Legal Information

Tough-Tags is a trademark of Diversified Biotech

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service