Key Documents
Z740256
Tough-Spots®
size 3/8 in., blue
Synonym(s):
solvent resistant labels, temperature resistant labels
About This Item
UNSPSC Code:
55121611
NACRES:
NB.35
material
PVC
blue
packaging
pkg of 1000 ea (per roll)
manufacturer/tradename
Diversified Biotech T-SPOTS-B
size
3/8 in.
General description
Tough - Chemically inert, polyolefin labels resist most organic solvents and caustic agents. They adhere to most plastics†, glass, and metals without cracking, peeling, or degrading
Temperature-Resistant - From -196°C to 100°C, Tough-Spots® withstand boiling water baths, moderate oven temperatures (150°C), autoclaving and extreme freezer temperatures (liquid and vapor phase nitrogen)
Specially Designed - Label sheets are specially top-coated to readily accept laser printing. Heat-resistant, stay-flat sheets will not jam in laser printers
Ideally Sized - Fits a variety of microcentrifuge tube tops, cryovial caps and lids, as well as many other laboratory container caps, lids and covers
Color Coding - 3/8 in. (9.5 mm) and 1/2 in. (13 mm) diameter Laser Tough-Spots® are available in White, Blue, Green, Orange, Red and Yellow
Legal Information
Tough-Spots is a registered trademark of Diversified Biotech