Key Documents
L5539
Sigma-Aldrich
Laser Tough-Spots®
diam. 3/8 in., green
Synonym(s):
solvent resistant labels, temperature resistant labels
About This Item
UNSPSC Code:
55121611
NACRES:
NB.35
material
green
polyolefin
packaging
pack of 3840 ea (192 labels per sheet)
manufacturer/tradename
Diversified Biotech SPOT-1200
diam.
3/8 in.
General description
Tough-Spots are chemically inert, polyvinyl labels that adhere to all plastics. Tough-Spots withstand boiling water baths, conventional and cryogenic freezer temperatures, most organic solvents, caustic agents and other challenges without peeling, becoming brittle or degrading. Labels can be printed directly from the laser printer. 20 sheets per pack.
Legal Information
Tough-Spots is a registered trademark of Diversified Biotech