Key Documents
Z719935
Sigma-Aldrich
Spinbar® Capsule Magnetic Stirring Bar
pkg of 1 ea
Synonym(s):
Stir Bar, Stirbar, Stirring Bar
About This Item
UNSPSC Code:
41103806
NACRES:
NB.72
material
polygon PTFE-coated (in flat housing)
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art F37152-0000
bar L × D
1.3 cm × 5.3 cm
General description
Designed for stirring in awkward vessels, for gently mixing staining solutions above delicate gels, or for quickly dissolving chemicals. A small flat capsule contains the magnetic stirring bar that spins freely inside without contact with the vessel bottom or gel. The protective capsule is molded with polystyrene and shelters a Spinbar magnetic stirring bar which is molded with inert PTFE. Bar diameter is 1 3/16 in.
Legal Information
Spinbar is a registered trademark of SP Scienceware, a division of SP