Documents
HS120547
Magnetic Stir Bar
L × diam. 10 mm × 5 mm
Synonym(s):
Magnetic Stirring Rod, Stir Bar Magnet
About This Item
UNSPSC Code:
41122401
NACRES:
NB.72
material
(with pivot ring)
octagonal
round white PTFE stir bar(s)
packaging
pack of 10 ea
manufacturer/tradename
Heathrow Scientific 120547
L × diam.
10 mm × 5 mm
General description
The ALNICO magnetic stir bar withstands temperatures up to 250°C, making them suitable for use with both unheated and heated stirrers. Low friction octagon shape with a center pivot ring helps create a strong mixing vortex, even with more viscous solutions
Features and Benefits
  • Magnetic bar is made from ALNICO, one of the strongest permanent magnets available
  • The sintered manufacturing process reduces the chance of decoupling (spin outs), which weaker stir bars often suffer from
  • Encapsulation in PTFE protects both magnet and what it is stirring
  • Chemically inert and easy to clean
  • Can withstand temperatures up to 250 °C, making them suitable for use with both unheated and heated stirrers
  • Low friction octagon shape with a center pivot ring helps create a strong mixing vortex, even with more viscous solutions