Z708194
Pipettor guide
aligns pipetter with the multiwell plates, pkg of 1 ea
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Recommended Products
packaging
pkg of 1 ea
manufacturer/tradename
Diversified Biotech
General description
Features a plastic sliding guide, above the multiwell plate to help align your pipetter with the multiwell plate wells. The sliding guide is clearly marked with column letters for fast and easy orientation. Compatible with multichannel pipetters; use with or without the 30° angled stand. A great tool to reduce eyestrain and tedious pipetting work.
Choose from one of the most recent versions:
Certificates of Analysis (COA)