Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z707538

TPP® tissue culture flasks

capacity 15 mL, Tissue Culture (TC)-treated surface, with cap (Filtered), case of 360 ea

Synonym(s):

T25 flask, cell culture flask, tissue culture flask

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122104
NACRES:
NB.22

material

angled neck
polystyrene
screw top cap

sterility

sterile

feature

with cap (Filtered)

packaging

pack of 10
case of 360 ea

manufacturer/tradename

TPP 90026

capacity

15 mL

external L × W × H

90 mm × 50 nm × 25 mm

surface area

25 cm2 , growth surface (cm2)

working volume

3-5 mL

suitability

suitable for (cell culture; tissue culture)

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

TPP Tissue Culture Flasks
Inside surface treatment by TPP provides an optimal growth surface on the flask base for the most varied matrix-dependent tissue cultures. The untreated side and top inside surfaces reduce the amount of cell loss during the separation phases, because undesired tissue adhesion outside the growth zone is diminished.

The flask′s innovative shape is of such a design that devices inserted through the flask′s neck have optimal access to the entire base surface. This means that there are no "dead" corners that cannot be reached with pipettes or cell scrapers. This excellent accessibility is due to the large neck opening and the favorable angle of inclination. The angled neck also reduces the chance of accidental spills and permits "dribble-free" pouring.

The TPP filter cap has a welded membrane in the cap cavity to offer free gas exchange without gas flow alteration. For optimal flask breathing, all membranes are proportionally dimensioned to each cap size. The microporous hydrophobic membrane (cytotoxic free) itself is constructed of 2 layers of membrane material based on a white acrylic copolymer over nylon support. With an airflow rate of 4.5 (LPM /3.7 cm2/ 13.5 psi), the gas exchange properties of the filtered flasks are comparable to that of the standard cap flasks.

The TPP tissue culture flasks stand for high quality aspects.

Advantages of the tissue culture flask:
  • Uniform even surface of the growth area
  • Crystal clear transparency
  • Stable, slip-free stacking of several flasks
  • Bilateral marking area
  • Accurate volume scale for optic control of the filling volume
  • Excellent access of pipettes, tissue scrapers and tissue spatula
  • Marking area with lines on both sides to allow your identification
  • Increased overflow height reduces the danger of slopover of media
  • "VENT" and filter screw caps

For optimum tissue growth, the growth area is opto-mechanical treated. This enhances cell adhesion and cell growth.

The geometry of the neck guarantees total access of TPP pipettes, TPP scrappers and the TPP spatula.
The inside surface treatment by TPP provides an optimal growth surface on the flask base for the most varied matrix-dependent tissue cultures. The untreated side and top inside surfaces reduce the amount of cell loss during the separation phases, because undesired tissue adhesion outside the growth zone is diminished. The flask′s innovative shape is of such a design that devices inserted through the flask′s neck have optimal access to the entire base surface. This means that there are no "dead" corners that cannot be reached with pipettes or cell scrapers.

This excellent accessibility is due to the large neck opening and the favorable angle of inclination. The angled neck also reduces the chance of accidental spills and permits "dribble-free" pouring.

When the vented screw cap is tightened to the end position, the closed tissue culture flask is liquid and gas tight. By simply unscrewing the cap one quarter of a turn, the flask is in the "VENT" position, with the inside of the flask being aerated through a defined gap opening. This guarantees that the gas exchange with the environment and the possible risk of contamination are kept to a minimum. In addition, the special flat thread profile of the caps stops excess tightening and thus allows the caps to be re-used multiple times.

The TPP filter cap has a welded membrane in the cap cavity to offer free gas exchange without gas flow alteration. For optimal flask breathing, all membranes are proportionally dimensioned to each cap size. The microporous hydrophobic membrane (cytotoxic free) itself is constructed of 2 layers of membrane material based on a white acrylic copolymer over nylon support. With an airflow rate of 4.5 (LPM/3.7cm2/13.5psi), the gas exchange properties of the filtered flasks are comparable to that of the standard cap flasks.

Legal Information

TPP is a registered trademark of TPP Techno Plastic Products AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service