Z688746
Nunc® IVF multidish
Δ surface treatment, 4 well dish, 1.9 cm2/well, 120/cs
Synonym(s):
4 well culture dish, 4 well multidish
About This Item
Recommended Products
material
polystyrene
square
sterility
sterile
sterile
feature
lid: no
packaging
case of 120 ea
manufacturer/tradename
Nunc 144444
dish L × W
66 mm × 66 mm
plate L × W
66 mm × 66 mm
size
4 wells
surface area
1.9 cm2 , per well
well surface area
1.9 cm2
color
clear
suitability
suitable for (in vitro fertilization)
binding type
Nunclon Delta surface
Looking for similar products? Visit Product Comparison Guide
General description
- Full batch control of all components in the final product for full traceability
- Unique certification based on a 1-cell stage mouse embryo toxicity test
- The certificate confirms: Sterility (SAL 10-6), Non-pyrogenicity, USP Class VI test
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service