Documents
Z376787
Sigma-Aldrich
Disposable culture tubes
polystyrene tube
Synonym(s):
culture tube, plastic test tube, test tubes
About This Item
UNSPSC Code:
41121706
NACRES:
NB.22
material
polystyrene tube
round bottom
straight neck
translucent tube
sterility
non-sterile
feature
RCF 1,400 × g
neck thread push fit
packaging
pack of 1000 ea
manufacturer/tradename
Simport T400-3
capacity
5 mL
size
12 mm × 75 mm
General description
Ideal for use in bacteriology, RIA, coagulation, and other routine laboratory procedures. Polypropylene tubes can be centrifuged at 3000 × g and sterilized at 120°C. Polystyrene tubes can be centrifuged at 1400 × g, but can not be autoclaved.