Key Documents
Z373443
Sigma-Aldrich
LiposoFast Liposome Factory
Replacement gas-tight 0.5 mL syringe
About This Item
UNSPSC Code:
41100000
NACRES:
NB.22
General description
Fast, efficient formation of uniform-sized, unilamellar liposomes. The principle is simple: a lipid emulsion is repeatedly extruded through a porous polycarbonate membrane, forced back and forth by specially modified gas-tight syringes attached to the membrane support capsule. With the syringes provided, unit has 0.5 mL capacity and virtually zero dead volume. Membranes from 100 to 400 nm pore size available.
Choose from one of the most recent versions:
Certificates of Analysis (COA)