Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z366218

Sigma-Aldrich

Cryo-Babies® Labels

Cryo-Babies® Labels

white, L × W 1.28 in. × 0.5 in., roll of 1000 labels

Synonym(s):

cryogenic vial labels, freezer vial labels

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
55121611
NACRES:
NB.35
Pricing and availability is not currently available.

material

(Polyvinyl)
white

packaging

roll of 1000 labels

manufacturer/tradename

Diversified Biotech CRY-1000

technique(s)

cryopreservation: suitable

L × W

1.28 in. × 0.5 in.

Legal Information

Cryo-Babies is a registered trademark of Diversified Biotech

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Are these laser printer sheets?

    1 answer
    1. These are blank labels that are used for hand-labeling cryogenic vials, microtubes, test tubes, glass vials, beakers, and metal racks. These are not suitable laser printers.

      Helpful?

Reviews

Active Filters

    • Review 1
    • Votes 0
    4 out of 5 stars.

    Good labels

    Easy to use labels you hand-write on. Need to use a decent pen. Labels can withstand low temperatures.

    Helpful?

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service