Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z359041

Sigma-Aldrich

Rack for Nalgene® cryogenic vials

Holds 25 vials in a staggered array

Synonym(s):

cryogenic vial storage, cryovial rack

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122806
NACRES:
NB.22

material

polycarbonate

feature

autoclavable
Holds 25 x vials

packaging

pack of 4 ea

manufacturer/tradename

Nalgene® 5030-0505

technique(s)

cryopreservation: suitable

color

blue

Looking for similar products? Visit Product Comparison Guide

General description

Wider spacing for easier manipulation
Accept all Nalgene cryogenic vials. Bottom of each well interlocks with vial, allowing one-hand manipulation.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service