Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS431119

Corning® cryogenic boxes, racks & trays

Box for 81 cryogenic vials, 1-2 mL

Synonym(s):

cryovials

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123304
NACRES:
NB.13

sterility

non-sterile

feature

Holds 81 x 1-2 mL tubes

packaging

pack of 10 ea

manufacturer/tradename

Corning 431119

technique(s)

cryopreservation: suitable

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Reusable orange polycarbonate cryogenic vial storage box with lid
  • Box can withstand temperature from - 196°C to +121°C
  • Grooves on the sides of the box improve grip when wearing gloves

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service