Key Documents
Z329398
BRAND® magnetic stirring bar, PTFE, triangular
L 55 mm, bar diam. 14 mm
Synonym(s):
PTFE, Stirring rods
About This Item
UNSPSC Code:
41103806
NACRES:
NB.72
material
triangular PTFE-coated
packaging
pack of 10
manufacturer/tradename
BRAND 137822
L
55 mm
bar diam.
14 mm
General description
Wedge shaped. Ideal for dissolving solids and mixing of sediments due to scraping action. Angular surface creates high turbulence even at low speeds.
Other Notes
BR137822 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG