Skip to Content
MilliporeSigma

Skip To

Z2000

β-Zearalenol

≥98%

Synonym(s):

beta-Zearalenol, 2,4-Dihydroxy-6-(6β,10-dihydroxy-trans-1-undecenyl)benzoic acid μ-lactone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C18H24O5
CAS Number:
Molecular Weight:
320.38
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352100
MDL number:
Assay:
≥98%
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98%

form

powder

storage temp.

−20°C

SMILES string

C[C@H]1CCC[C@@H](O)CCC\C=C\c2cc(O)cc(O)c2C(=O)O1

InChI

1S/C18H24O5/c1-12-6-5-9-14(19)8-4-2-3-7-13-10-15(20)11-16(21)17(13)18(22)23-12/h3,7,10-12,14,19-21H,2,4-6,8-9H2,1H3/b7-3+/t12-,14-/m0/s1

InChI key

FPQFYIAXQDXNOR-PMRAARRBSA-N

Gene Information

rat ... Ar(24208)

Application

Reactant involved in biological studies including:
  • Synthesis of amino glycoside nucleotides
  • Enzymatic synthesis of glucuronides of zearalenone
  • Derivatization of Zearalenone

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Z0166Z0417Z0167
assay

≥98%

assay

≥98% (HPLC)

assay

~98% (HPLC)

assay

≥98% (TLC)

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

powder

form

powder

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

Gene Information

rat ... Ar(24208)

Gene Information

rat ... Ar(24208)

Gene Information

rat ... Ar(24208)

Gene Information

rat ... Ar(24208)


pictograms

Health hazardCorrosion

signalword

Danger

Hazard Classifications

Repr. 2 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters