Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z0166

Sigma-Aldrich

α-Zearalenol

Synonym(s):

2,4-Dihydroxy-6-(6α,10-dihydroxy-trans-1-undecenyl)benzoic acid μ-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24O5
CAS Number:
Molecular Weight:
320.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

solubility

methanol: 9.80-10.20 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

C[C@H]1CCC[C@H](O)CCC\C=C\c2cc(O)cc(O)c2C(=O)O1

InChI

1S/C18H24O5/c1-12-6-5-9-14(19)8-4-2-3-7-13-10-15(20)11-16(21)17(13)18(22)23-12/h3,7,10-12,14,19-21H,2,4-6,8-9H2,1H3/b7-3+/t12-,14+/m0/s1

InChI key

FPQFYIAXQDXNOR-QDKLYSGJSA-N

Gene Information

rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Application

α-Zearalenol has been used:
  • as an analytical standard in the quantification of mycotoxins from animal feed
  • to evaluate its oestrogenic activity on boar sperm motility , as an anti-oestrogen control in MCF7 cells
  • as a reference standard in a liquid chromatography-negative ion electrospray tandem mass spectrometry with atmospheric pressure chemical ionization (LCAPCI/MS/MS) for bovine follicular fluids (FFs) quantification

Biochem/physiol Actions

α-Zearalenol (α-ZOL) is a catabolite of zearalenone, synthesized majorly in granulosa cells, intestine and liver in pigs. It has estrogenic functionality. α-ZOL inhibits sperm motility in horse possibly by eliciting genotoxic effect. α-ZOL displays higher affinity towards estrogenic receptors.

Pictograms

CorrosionHealth hazard

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Repr. 2 - Skin Corr. 1B - STOT SE 2

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Zearalenone and its derivatives alpha-zearalenol and beta-zearalenol decontamination by Saccharomyces cerevisiae strains isolated from bovine forage
Keller L, et al.
Toxins, 7(8), 3297-3308 (2015)
Dose-response effects of estrogenic mycotoxins (zearalenone, alpha-and beta-zearalenol) on motility, hyperactivation and the acrosome reaction of stallion sperm
Filannino A, et al.
Reproductive Biology and Endocrinology, 9(1), 134-134 (2011)
Effects of zearalenone, alpha-zearalenol, and genistein on boar sperm motility in vitro
Krejcarkova A, et al.
Czech Journal of Animal Science, 62(10), 435-445 (2017)
Detection of zearalenone and its metabolites in naturally contaminated follicular fluids by using LC/MS/MS and in vitro effects of zearalenone on oocyte maturation in cattle
Takagi M, et al,
Reproductive Toxicology, 26(2), 164-169 (2008)
Oestrogenic potencies of Zeranol, oestradiol, diethylstilboestrol, Bisphenol-A and genistein: implications for exposure assessment of potential endocrine disrupters
Leffers H, et al.
Human Reproduction, 16(5), 1037-1045 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service