Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

X3004

Sigma-Aldrich

XAV939

≥98% (HPLC), powder, tankyrase inhibitor

Synonym(s):

3,5,7,8-Tetrahydro-2-[4-(trifluoromethyl)phenyl]-4H-thiopyrano[4,3-d]pyrimidin-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11F3N2OS
CAS Number:
Molecular Weight:
312.31
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

XAV939, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear

storage temp.

room temp

SMILES string

FC(F)(F)c1ccc(cc1)C2=NC3=C(CSCC3)C(=O)N2

InChI

1S/C14H11F3N2OS/c15-14(16,17)9-3-1-8(2-4-9)12-18-11-5-6-21-7-10(11)13(20)19-12/h1-4H,5-7H2,(H,18,19,20)

InChI key

KLGQSVMIPOVQAX-UHFFFAOYSA-N

Application

XAV939 has been used in the inhibition
  • of tankyrase in neuroblastoma cell lines
  • of diphtheria toxin-like ADP-ribosyltransferases (ARTD) in medulloblastoma cells
  • of wnt signalling in human induced pluripotent stem cells (hiPSCs)

Biochem/physiol Actions

XAV939 is a Tankyrase inhibitor (thereby inhibiting Wnt / β-catenin signaling)
XAV939 is a cell-permeable dihydrothiopyranopyrimidinol, which elicits anti-tumor functionality against colon, lung, breast and liver cancer. It binds to catalytic poly-ADP-ribose polymerase (PARP) domain of the tankyrase enzyme resulting in β-catenin destruction. It also inhibits tankyrase in neuroblastoma promoting apoptosis in tumor cells.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Other Notes

XAV939 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the XAV939 probe summary on the Chemical Probes Portal website.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robust generation of cardiomyocytes from human iPS cells requires precise modulation of BMP and WNT signaling
Kadari A, et al.
Stem Cell Reviews and Reports, 11(4), 560-569 (2015)
Tankyrase inhibitors attenuate WNTbeta-catenin signaling and inhibit growth of hepatocellular carcinoma cells
Ma L, et al.
Testing, 6(28), 25390-25390 (2015)
Jing Sun et al.
Cancer medicine, 6(4), 834-844 (2017-03-24)
Golgi phosphoprotein 3 (GOLPH3), a newly recognized oncogene, is associated with tumor growth, metastasis, and poor prognosis in several types of cancer. However, its biological role and underlying mechanism in epithelial ovarian cancer (EOC) remain poorly understood. Here, we found
XAV939, a tankyrase 1 inhibitor, promotes cell apoptosis in neuroblastoma cell lines by inhibiting Wntbeta-catenin signaling pathway
Tian XH, et al.
Journal of Experimental & Clinical Cancer Research, 32(1), 100-100 (2013)
XAV939-mediated ARTD activity inhibition in human MB cell lines
Renna C, et al.
Testing, 10(4), e0124149-e0124149 (2015)

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service