Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

WH0001486M1

Sigma-Aldrich

Monoclonal Anti-CTBS antibody produced in mouse

clone 1B5-1B9, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CTB, Anti-chitobiase, di-N-acetyl-

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1B5-1B9, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CTBS(1486)

General description

The gene CTBS (chitobiase) is mapped to human chromosome 1p22. The gene spans a length of approximately 20kb with seven exons interspaced by six introns. The 5′-flanking region of human chitobiase gene is rich in GC-content.

Immunogen

CTBS (AAH24007.2, 37 a.a. ~ 105 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
DCPCPEPELCRPIRHHPDFEVFVFDVGQKTWKSYDWSQITTVATFGKYDSELMCYAHSKGARVVLKGNL

Biochem/physiol Actions

The gene CTBS (chitobiase) encodes a lysosomal glycosidase that functions in the degradation of asparagine-linked glycoproteins. It cleaves the reducing-end N-Acetylglucosamine residues from the chitobiose core of oligosaccharides.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure of the human gene for lysosomal di-N-acetylchitobiase.
Liu B
Glycobiology, 9, 589-593 (1999)
Lysosomal chitobiase (CTB) and the G-protein gamma 5 subunit (GNG5) genes co-localize to human chromosome 1p22.
Ahmad W
Cytogenetics and Cell Genetics, 71, 44-46 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service