Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07-2126

Sigma-Aldrich

Anti-phospho-FOXO1 Antibody (Thr24)

from rabbit, purified by affinity chromatography

Synonym(s):

Forkhead box protein O1, Forkhead box protein O1A, Forkhead in rhabdomyosarcoma

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rabbit

Quality Level

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pThr24)

Gene Information

human ... FOXO1(2308)

General description

Forkhead box O (FOXO) transcription factors play an important role in modulating metabolic functions and cell proliferation. Given the relatively high expression of FOXO1 in insulin-responsive tissues, this transcription factor is highly poised to regulate energy metabolism. When nutrient and insulin levels are low, FOXO1 promotes expression of gluconeogenic enzymes. Conversely, in the fed state, insulin levels rise and stimulate uptake of glucose primarily into skeletal muscle and other organs, including adipose tissue. Under certain pathophysiologic conditions, including insulin resistance, negative signaling to FOXO1 is compromised. Increased cell proliferation occurs when FOXO1 is inactivated when phosphorylated by Akt at Thr24.

Specificity

This antibody recognizes FOXO1 phosphorylated at Thr24.

Immunogen

Epitope: Phosphorylated Thr24
KLH-conjugated linear peptide corresponding to human FOXO1 phosphorylated at Thr24.

Application

Research Category
Signaling
Research Sub Category
Transcription Factors
Use Anti-phospho-FOXO1 (Thr24) Antibody (Rabbit Polyclonal Antibody) validated in WB to detect phospho-FOXO1 (Thr24) also known as Forkhead box protein O1.

Quality

Evaluated by Western Blot in HEK293 cell lysate untreated and BMP2-treated.

Western Blot Analysis: 2 µg/mL of this antibody detected phospho-FOXO1 (Thr24) in HEK293 cell lysate untreated and BMP2-treated.

Target description

~78 kDa observed.
Calculated molecular weight is 70 kDa FOXO1 has been shown to migrate to ~78 kDa in western blots (Kamei, Y., et al. (2004). JBC. 279(39):41114-41123).

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HEK293 cell lysate untreated and BMP2-treated

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service