Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V5515

Sigma-Aldrich

Porcine Brain Membrane Homogenate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.56

biological source

Porcine

Quality Level

description

A source of Saxitoxin (STX) receptors

form

suspension

concentration

8-15 mg protein/mL (BCA)

technique(s)

ligand binding assay: suitable

shipped in

dry ice

storage temp.

−70°C

Application

Paralytic shellfish poisoning results from toxins originating primarily from algae of the genus Alexandrium causing intoxication and significant health risk for human consumers. This sample of porcine brain membrane homogenate is developed for use in receptor binding assays for determination of paralytic shellfish poisoning as described by Van Dolah et al. This assay provides a high throughput method for screening for paralytic shellfish poisoning and study of the receptors/toxins involved.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Single-Laboratory Validation of the Microplate Receptor Binding Assay for Paralytic Shellfish Toxins in Shellfish
Van Dolah, F. M., Leighfield, T. A., Doucette, G. J.
Journal of AOAC (Association of Official Analytical Chemists) International, 92, 1706-1713 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service