Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T9661

Sigma-Aldrich

Eppendorf® Safe-Lock microcentrifuge tubes

volume 1.5 mL, natural

Synonym(s):

eppendorf tubes, microcentrifuge tubes, microfuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
NACRES:
NB.11

material

cap (Safe-Lock lid)
polypropylene

sterility

non-sterile

feature

RCF 30,000 × g

manufacturer/tradename

Eppendorf® 0030 120.086 (EU)
Eppendorf® 022363204 (USA)

volume

1.5 mL

color

natural

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Our Safe Lock microtubes are designed to offer excellent centrifugation stability and optimized sealing properties and ensure your valuable samples remain safe and secure.

Application

Eppendorf® Safe-Lock microcentrifuge tubes are suitable for DNA recovery from plasmids.

Features and Benefits

  • The hinged Eppendorf Safe-Lock lid prevents the opening of the lid during incubation and storage
  • The tubes are devoid of slip agents, plasticizers, and biocides
  • The polypropylene tubes are highly resistant to chemicals, mechanical stress, and high temperatures
  • The lid sealing prevents higher evaporation during storage
  • Large frosted lid and surface on the side for easy labeling
  • Functionality varies from -86 °C to 100 °C
  • When open, it can be autoclaved at 121 °C for 20 mins
  • Exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and ensures safety while working with hazardous samples
Tubes snap closed and stay closed. Flat top with writing surface for positive sample identification

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

GenomePlex® Whole Genome Amplification has been used to amplify genomic DNA from soybean, corn, tomato, purple coneflower, and ginseng.

Techniques for protein antigen molecular weight determination, protein interactions, enzymatic activity, and post-translational modifications.

Techniques for protein antigen molecular weight determination, protein interactions, enzymatic activity, and post-translational modifications.

Techniques for protein antigen molecular weight determination, protein interactions, enzymatic activity, and post-translational modifications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service