Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP0030119401

Eppendorf Tube®

capacity 5.0 mL, Eppendorf Quality, pack of 200 ea (2 bags of 100 ea)

Synonym(s):

eppendorf tubes, microcentrifuge tubes, microfuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.11

material

cap (snap cap)
colorless cap
colorless tube
conical bottom
polypropylene cap
polypropylene tube
transparent polypropylene

sterility

sterile

feature

graduations
Eppendorf Quality
conical shaped bottom with hinged lid

packaging

pack of 200 ea (2 bags of 100 ea)

manufacturer/tradename

Eppendorf® 0030119401

parameter

25,000 × g

capacity

5.0 mL

Looking for similar products? Visit Product Comparison Guide

General description

Eppendorf Tubes are the perfect option for working with medium-sized sample volumes!

  • Maximum safety and stability for centrifugation up to 25,000 × g
  • Conical bottoms
  • Easier processing of samples up to 5.0 mL
  • Higher yields in DNA isolation, prep of mastermixes and buffers, and safe cell and tissue lysis
  • Transparent polypropylene, free of plasticizers, biocides or mold release agents
  • Hinged lid for minimized sample evaporation during storage and incubation in a wide range of temperatures from -86°C to 80°C

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Quality is a trademark of Eppendorf AG
Eppendorf Tube is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service