Skip to Content
MilliporeSigma
All Photos(1)

Documents

T3160

Sigma-Aldrich

TC-100 Insect Medium

With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for insect cell culture

Synonym(s):

BML-TC/10

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | insect: suitable

components

glucose: 1.0 g/L
L-glutamine: 0.6 g/L
NaHCO3: 0.35 g/L

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

TC-100 Insect Medium has been developed to optimize the production of Autographica californica nuclear polyhedrosis virus (NPV) virions by cells from the fall armyworm, Spodoptera frugiperda. It has been found to support the growth of cells derived from a variety of lepidopteran species. The medium is mostly employed in the growth of baculoviruses in lepidopteran cell lines.

Application

For baculovirus culturing in lepidoptera.
TC-100 Insect Medium has been used:
  • in the transfection of insect cells in suspension for efficient baculovirus generation
  • to harvest Varroa destructor saliva
  • as a starter media to culture and maintain insect cells such as Spodoptera frugiperda

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Roest et al.
MethodsX, 3, 371-377 (2016-05-26)
Baculovirus (BV) mediated insect cell expression system utilizes transfection as a first step to introduce recombinant baculovirus DNA into insect cells. Many labs are still relying on the conventional liposome based transfection method in adherent culture. Here we describe a
Insect Biotechnology
Short Views on Insect Genomics and Proteomics, 185-210 (2016)
Yi Zhang et al.
Journal of economic entomology, 112(2), 505-514 (2018-09-17)
The Varroa destructor (Acari Varroidae) mite is a serious threat to honey bee due to hemolymph feeding and virus transmission. Mite salivary proteins are involved in these interactions. However, the salivary secretome has not been previously characterized. In this paper

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service