Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T2663

Sigma-Aldrich

Trizma® hydrochloride solution

1 M, BioReagent, for molecular biology

Synonym(s):

Tris hydrochloride solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

grade

Molecular Biology
for molecular biology
for molecular biology

Quality Level

sterility

0.2 μm filtered

product line

BioReagent

form

solution

concentration

1 M

impurities

DNase, RNase, NICKase and protease, none detected

pH

7.35-7.45

SMILES string

Cl.NC(CO)(CO)CO

InChI

1S/C4H11NO3.ClH/c5-4(1-6,2-7)3-8;/h6-8H,1-3,5H2;1H

InChI key

QKNYBSVHEMOAJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trizma® hydrochloride solution is a useful biological buffer.

Application

The pH values of all buffers are temperature and concentration dependent. For Tris buffers, pH increases about 0.03 unit per degree C decrease in temperature, and decreases 0.03-0.05 unit per ten-fold dilution.
For precise applications, use a carefully calibrated pH meter with a glass/calomel combination electrode.
Trizma® hydrochloride along with Trizma-base is used to prepare buffer solutions of pH 7.35. It was used to prepare the triton extract. It may be used in the analysis of protease activity, isolated from the Bacillus stearothermophilus and Bacillus subtilis. It may be employed as buffer for the restriction fragment analysis of genomic DNA isolated from the enterococcal isolates and Escherichia coli.

Other Notes

Prepared with pH-adjusted Biotechnology Performance Certified Trizma Base in 18 megohm water and 0.2 μm filtered.

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B E Murray et al.
Journal of clinical microbiology, 28(9), 2059-2063 (1990-09-01)
Epidemiologic evaluation of enterococci has been limited by the lack of a simple and effective method for comparing strains. In this study, we have compared chromosomal restriction endonuclease digestion patterns of 27 isolates of Enterococcus faecalis from three different locations
M Fujii et al.
Journal of bacteriology, 154(2), 831-837 (1983-05-01)
The structural gene for a thermostable protease from Bacillus stearothermophilus was cloned in plasmid pTB90. It is expressed in both B. stearothermophilus and Bacillus subtilis. B. stearothermophilus carrying the recombinant plasmid produced about 15-fold more protease (310 U/mg of cell
Reconstitution and purification of the D-glucose transporter from human erythrocytes.
M Kasahara et al.
The Journal of biological chemistry, 252(20), 7384-7390 (1977-10-25)
El-Refaie Kenawy et al.
Journal of controlled release : official journal of the Controlled Release Society, 81(1-2), 57-64 (2002-05-07)
Electrospun fiber mats are explored as drug delivery vehicles using tetracycline hydrochloride as a model drug. The mats were made either from poly(lactic acid) (PLA), poly(ethylene-co-vinyl acetate) (PEVA), or from a 50:50 blend of the two. The fibers were electrospun
Asad Jan et al.
Nature protocols, 5(6), 1186-1209 (2010-06-12)
The amyloid cascade hypothesis, supported by strong evidence from genetics, pathology and studies using animal models, implicates amyloid-beta (Abeta) oligomerization and fibrillogenesis as central causative events in the pathogenesis of Alzheimer's disease (AD). Today, significant efforts in academia, biotechnology and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service