Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
CAS Number:
UNSPSC Code:
12352202
EC Number:
234-318-8
NACRES:
NA.61
MDL number:
eCl@ss:
34058011
Form:
powder
Assay:
≥98%
Biological source:
bovine
Pricing and availability is not currently available.
biological source
bovine
Quality Segment
assay
≥98%
form
powder
technique(s)
cell culture | mammalian: suitable
solubility
H2O: 20 mg/mL
UniProt accession no.
storage temp.
2-8°C
Gene Information
bovine ... TF(280705)
Application
All cells require iron for the proper uptake of oxygen from their environment. Transferrin, a protein isolated from serum, causes the uptake and transport of iron from culture medium to the cells. This allows proper oxygen uptake and also stimulates growth-related enzyme activity. Iron-saturated transferrin (holo transferrin) is continually recycled after releasing iron and becomes iron-deficient transferrin (apo-transferrin). Choosing which form to use depends upon the culture conditions. In media such as Dulbecco′s Modified Eagle′s Medium/Ham′s Nutrient Mixture F12 (DMEM/F12) that contain high levels of iron salt, using apo-transferrin is preferred.
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥98% | assay ≥98% | assay ≥98% | assay ≥95% protein basis (biuret), ≥98% (agarose gel electrophoresis) |
| biological source bovine | biological source mouse | biological source human | biological source human |
| technique(s) cell culture | mammalian: suitable | technique(s) protein quantification: suitable | technique(s) cell culture | stem cell: suitable | technique(s) cell culture | mammalian: suitable |
| form powder | form powder | form powder | form powder |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
| Gene Information bovine ... TF(280705) | Gene Information mouse ... Trf(22041) | Gene Information human ... TF(7018) | Gene Information human ... TF(7018) |
Still not finding the right product?
Explore all of our products under apo-Transferrin bovine
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



