Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SYNB140100

Synthware Glass wide mouth media bottle

borosilicate glass 3.3, capacity 100 mL, green polypropylene cap

Synonym(s):

Synthware B140100, glass graduated bottle with screw cap

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104812
NACRES:
NB.43

material

borosilicate glass 3.3
clear bottle
green polypropylene cap
wide-mouth

feature

with cap

packaging

pack of 1 ea

manufacturer/tradename

Kemtech B140250

capacity

100 mL

Looking for similar products? Visit Product Comparison Guide

General description

GL45 Wide Mouth Media Storage Bottle With Color Caps, 100mL includes Accessories: GL45-L

Application

kemetch; synthware; media bottle; GL45

Features and Benefits

Organic Chemistry

Legal Information

Synthware is a trademark of Kemtech America Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service