Skip to Content
MilliporeSigma
All Photos(1)

Documents

SRP6112

Sigma-Aldrich

G6PD from E. coli

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

Glucose-6-phosphate 1-dehydrogenase, Zwf

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.32

biological source

Escherichia coli

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

liquid

mol wt

55.7 kDa (491 aa, 1-491 aa)

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

Escherichia coli ... ZWF(7149716)

General description

Glucose-6-phosphate dehydrogenase (G6PD) is the rate-limiting enzyme of the pentose phosphate pathway, a metabolic pathway that supplies reducing energy to cells by maintaining the level of NADPH.
Specific activity is 8-10 units/ml obtained by measuring the increase of NADPH in absorbance at 340 nm resulting from the reduction of NAD or NADP.

Biochem/physiol Actions

Glucose-6-phosphate dehydrogenase (G6PD) converts glucose-6-phosphate into 6-phosphoglucono-delta-lactone and simultaneously produces NADPH. The NADPH in turn maintains the level of glutathione in these cells that helps protect the red blood cells against oxidative damage. G6PD deficiency causes acute hemolytic anemia.

Physical form

1 mg/mL solution in 50 mM MES pH 6.0, 0.1 mM PMSF, 2 mM EDTA, 0.5 mM DTT and 10% glycerol.

Preparation Note

Centrifuge the vial prior to opening.

Other Notes

MAVTQTAQAC DLVIFGAKGD LARRKLLPSL YQLEKAGQLN PDTRIIGVGR ADWDKAAYTK VVREALETFM KETIDEGLWD TLSARLDFCN LDVNDTAAFS RLGAMLDQKN RITINYFAMP PSTFGAICKG LGEAKLNAKP ARVVMEKPLG TSLATSQEIN DQVGEYFEEC QVYRIDHYLG KETVLNLLAL RFANSLFVNN WDNRTIDHVE ITVAEEVGIE GRWGYFDKAG QMRDMIQNHL LQILCMIAMS PPSDLSADSI RDEKVKVLKS LRRIDRSNVR EKTVRGQYTA GFAQGKKVPG YLEEEGANKS SNTETFVAIR VDIDNWRWAG VPFYLRTGKR LPTKCSEVVV YFKTPELNLF KESWQDLPQN KLTIRLQPDE GVDIQVLNKV PGLDHKHNLQ ITKLDLSYSE TFNQTHLADA YERLLLETMR GIQALFVRRD EVEEAWKWVD SITEAWAMDN DAPKPYQAGT WGPVASVAMI TRDGRSWNEF E

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert C Stanton
IUBMB life, 64(5), 362-369 (2012-03-21)
Glucose-6-phosphate dehydrogenase (G6PD) is the rate-limiting enzyme of the pentose phosphate pathway. Many scientists think that the roles and regulation of G6PD in physiology and pathophysiology have been well established as the enzyme was first identified 80 years ago. And
T J Vulliamy et al.
Proceedings of the National Academy of Sciences of the United States of America, 85(14), 5171-5175 (1988-07-01)
Glucose-6-phosphate dehydrogenase (G6PD; EC 1.1.1.49) deficiency is a common genetic abnormality affecting an estimated 400 million people worldwide. Clinical and biochemical analyses have identified many variants exhibiting a range of phenotypes, which have been well characterized from the hematological point

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service