Skip to Content
MilliporeSigma

346774

Glucose-6-phosphate Dehydrogenase, Leuconostoc mesenteroides

Catalyzes the conversion of glucose-6-phosphate to 6-phosphogluconate.

Synonym(s):

Glucose-6-phosphate Dehydrogenase, Leuconostoc mesenteroides

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
EC Number:
UNSPSC Code:
12352202
NACRES:
NA.21
MDL number:
Specific activity:
≥200 units/mg protein
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

lyophilized

specific activity

≥200 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

foreign activity

Creatine phosphokinase ≤0.002%, Phosphohexose isomerase ≤0.02%

shipped in

ambient

storage temp.

−20°C

General description

Native glucose-6-phosphate dehydrogenase from Leuconostoc mesenteroides. Catalyzes the conversion of glucose-6-phosphate to 6-phosphogluconate. The enzyme can use either NAD+ or NADP+ as an electron acceptor. Reaction velocity with NAD+ is ~1.8 times greater than that with NADP+. Note: 10,000 NADP units are equivalent to ~18,000 NAD units.

Preparation Note

Reconstitute to 1 mg/ml in 5 mM glycine buffer, pH 8.0. Agitation of solution may cause precipitation.

Other Notes

Levy, H.R. 1989. Biochem. Soc. Trans. 17, 313.
One unit is defined as the amount of enzyme that will reduce 1.0 µmol NADP per min at 30°C, pH 7.8.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library