Skip to Content
MilliporeSigma
All Photos(1)

Documents

SRP5205

Sigma-Aldrich

MBP, GST tagged human

recombinant, expressed in E. coli, ≥65% (SDS-PAGE), buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥65% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~36 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MBP(4155)

General description

Myelin basic protein (MBP), an important constituent of the myelin sheath of CNS (central nervous system), is an autoimmunogen. It is mapped to human chromosome 18q.

Biochem/physiol Actions

MBP or Myelin basic protein forms dimers across the cytoplasmic apposition during the formation of myelin. The region of the protein involved in folding, polymerization and substrate specificities is conserved in various species and it may have a specialized role in protein-lipid interactions in the myelin membrane. MBP is an efficient substrate for numerous protein kinases and share similarities with the MARCKS protein in terms of having extended conformations regulated by their environment, N-terminal modifications, a dual nature of interactions with lipids, and binding to actin and Ca2+-calmodulin. Myelin basic protein (MBP) is expected to play a vital role in demyelinating diseases such as MS (multiple sclerosis).

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B E Chapman et al.
Advances in experimental medicine and biology, 100, 207-220 (1978-01-01)
High resolution 13C and 1H NMR spectra of myelin basic protein over a range of pH and concentrations indicate that intramolecular folding of the polypeptide chain occurs in aqueous solution in the region of residues 85 to 116. At pH
G Harauz et al.
Molecular and cellular biochemistry, 209(1-2), 155-163 (2000-08-15)
Myelin basic protein (MBP) and myristoylated alanine-rich C-kinase substrate (MARCKS) are similar in terms of having extended conformations regulated by their environment (i.e., solubilised or lipid-associated), N-terminal modifications, a dual nature of interactions with lipids, binding to actin and Ca2+-calmodulin
Molecular characterization of a patient with central nervous system dysmyelination and cryptic unbalanced translocation between chromosomes 4q and 18q
Gunn S R, et al.
American Journal of Medical Genetics. Part A, 120(1), 127-135 (2003)
Structure and developmental regulation of Golli-mbp, a 105-kilobase gene that encompasses the myelin basic protein gene and is expressed in cells in the oligodendrocyte lineage in the brain.
Campagnoni AT, et al.
Test, 268(7), 4930-4938 (1993)
Variation within MBP gene predicts disease course in multiple sclerosis
Zhou Y, et al.
Brain and Behavior, 7(4), e00670-e00670 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service