Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5112

Sigma-Aldrich

p63, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

AIS, B(p51A), B(p51B), EEC3, KET, LMS, NBP, OFC8, RHS, SHFM4, TP53CP, TP53L, TP63, TP73L, p40, p51, p53CP, p73H, p73L

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~92 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TP63(8626)

General description

p63 is the primordial member of the p53 family that acts in a conserved process of monitoring the integrity of the female germline, whereas the functions of p53 are restricted to vertebrate somatic cells for tumor suppression. p63 protein plays an important role in the development and maintenance of stratified epithelial tissues. p63 is critical for maintaining the progenitor-cell populations that are necessary to sustain epithelial development and morphogenesis. Mutations in p63 are associated with ectodermal dysplasia, cleft lip/palate syndrome 3 (EEC3) and split-hand/foot malformation 4 (SHFM4).

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eun-Kyung Suh et al.
Nature, 444(7119), 624-628 (2006-11-24)
Meiosis in the female germ line of mammals is distinguished by a prolonged arrest in prophase of meiosis I between homologous chromosome recombination and ovulation. How DNA damage is detected in these arrested oocytes is poorly understood, but it is
A Yang et al.
Nature, 398(6729), 714-718 (1999-05-05)
The p63 gene, a homologue of the tumour-suppressor p53, is highly expressed in the basal or progenitor layers of many epithelial tissues. Here we report that mice homozygous for a disrupted p63 gene have major defects in their limb, craniofacial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service