Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABT11

Sigma-Aldrich

Anti-ZO-1 Antibody, clone R40.76

clone R40.76, from rat

Synonym(s):

tight junction protein 1, zonula occludens 1 protein, Zona occludens protein 1, Zonula occludens protein 1, zona occludens 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

R40.76, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Tjp1(21872)

General description

Tight junctions are complexes of proteins that create intercellular boundaries between the plasma membrane domains of epithelial and endothelial cells. Many of the tight junction-associated proteins are members of the membrane associated guanylate kinase (MAGUK) family and include occludin and zona occludin family members ZO-1, ZO-2 and ZO-3. These proteins are thought to have both structural and signaling roles, and are characteristically defined by three protein-protein interaction modules: the PDZ domain, the SH3 domain and the guanylate kinase (GuK) domain. ZO-1 forms complexes with either ZO-2 or ZO-3. In addition, these proteins can also associate with claudin, occludin and F-actin, at tight junction stands, where they provide a linkage between the actin cytoskeleton and the tight junction. ZO-1 expression is significantly reduced in many breast cancer lines. ZO-2 and ZO-3 are ubiquitously expressed within epithelial tight junctions, and unlike ZO-1, which is also expressed at cell junctions of cardiac myocytes, ZO-2 is not expressed in nonepithelial tissue.

Specificity

The antibody recognizes ZO-1.

Immunogen

Epitope: Unknown
Purified protein corresponding to mouse ZO-1.

Application

Anti-ZO-1 Antibody, clone R40.76 detects level of ZO-1 & has been published & validated for use in WB, IP, IH.
Immunofluorescence Analysis: A previous lot was used by an indepenedent laboratory in IF (D′Angelo Siliciano, 1988).
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Quality

Evaluated by Western blot in mouse brain lysate.

Western blot Analysis: 0.5 µg/mL of this antibody detected ZO-1 in 10 µg of mouse brain lysate.

Target description

~220 kDa

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse brain lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zaifeng Yi et al.
Molecular medicine reports, 12(3), 3615-3620 (2015-06-06)
Adrenomedullin (AM) is a pivotal endogenous vasoactive peptide, which can maintain epithelial barrier function in inflammatory bowel disease. Myosin light chain kinase (MLCK)‑dependent phosphorylated myosin light chain kinase (p‑MLC) is a key regulator of intestinal barrier function. The aim of
László Homolya et al.
PloS one, 9(3), e91921-e91921 (2014-03-20)
Polarization of hepatocytes is manifested by bile canalicular network formation and activation of LKB1 and AMPK, which control cellular energy metabolism. The bile acid, taurocholate, also regulates development of the canalicular network through activation of AMPK. In the present study
Beatriz Fernández-Gil et al.
PloS one, 12(4), e0174474-e0174474 (2017-04-14)
Radiotherapy-induced gut toxicity is among the most prevalent dose-limiting toxicities following radiotherapy. Prevention of radiation enteropathy requires protection of the small intestine. However, despite the prevalence and burden of this pathology, there are currently no effective treatments for radiotherapy-induced gut
Nermin M Kady et al.
Diabetes, 67(4), 769-781 (2018-01-25)
Tight junctions (TJs) involve close apposition of transmembrane proteins between cells. Although TJ proteins have been studied in detail, the role of lipids is largely unknown. We addressed the role of very long-chain (VLC ≥26) ceramides in TJs using diabetes-induced
Qitao Zhang et al.
Experimental eye research, 178, 212-222 (2018-10-20)
The daily shedding and renewal of photoreceptor outer segments (OS) is critical for maintaining vision. This process relies on the efficient uptake, degradation, and sorting of shed OS material by the retinal pigment epithelium (RPE). Poor OS degradation has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service