Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4135

Sigma-Aldrich

IL-3 from mouse

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

HCGF P-cell stimulation factorIL-3 MGC79398, Interleukin-3, MCGF (Mast cell growth factor)Multi-CSF, MGC79399

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

mouse

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

<0.05 ng/mL ED50

mol wt

~15.1 kDa

packaging

pkg of 10 μg

impurities

endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Il3(16187)

Looking for similar products? Visit Product Comparison Guide

General description

IL3 (interleukin 3) is an immunomodulatory cytokine and a multilineage hematopoietic growth factor. It is predominantly produced by activated CD4+ T cells and mast cells. Recombinant mouse IL-3 is a 15.1 kDa protein containing 135 amino acid residues.

Biochem/physiol Actions

IL3 (interleukin 3) was originally known as multicolony-stimulating factor due to its capability to promote the development of a wide variety of hematopoietic progenitor cells from bone marrow, such as basophils, neutrophils, eosinophils, erythrocytes, megakaryocytes, mast cells, macrophages, and dendritic cells. In vitro this protein facilitates murine hematopoietic progenitor cell differentiation, and the survival and activation of mature myeloid cells. It promotes the release of histamine and mast cell growth, differentiation, and mediator release, and hence, is implicated in allergic inflammation. It exhibits bi-functionality during osteoclastogenesis where it stimulates the development of osteoclast progenitors but suppresses the osteoclastogenic process. Certain variants in this gene confer protection against malaria attack, and thus, this gene might play a role in the pathophysiology of falciparum malaria.

Physical form

Sterile filtered and lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1 –1.0 mg/mL. The solution can then be diluted into other aqueous buffers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

IL3 variant on chromosomal region 5q31-33 and protection from recurrent malaria attacks.
Meyer CG, et al.
Human Molecular Genetics, 20(6), 1173-1181 (2011)
Interleukin-3-deficient mice have increased resistance to blood-stage malaria.
Auclair SR et al
Infection and Immunity, 82(3), 1308-1314 (2014)
Jing Wang et al.
EMBO molecular medicine, 6(7), 952-969 (2014-06-26)
Immunoglobulin E (IgE) activates mast cells (MCs). It remains unknown whether IgE also activates other inflammatory cells, and contributes to the pathogenesis of abdominal aortic aneurysms (AAAs). This study demonstrates that CD4+ T cells express IgE receptor FcεR1, at much
Serena De Vita et al.
PloS one, 9(5), e96209-e96209 (2014-05-03)
Systemic Mastocytosis (SM) is a clonal disease characterized by abnormal accumulation of mast cells in multiple organs. Clinical presentations of the disease vary widely from indolent to aggressive forms, and to the exceedingly rare mast cell leukemia. Current treatment of
Huixian Hong et al.
Biochemical and biophysical research communications, 440(4), 545-550 (2013-10-10)
Interleukin (IL)-3, a multilineage hematopoietic growth factor, is implicated in the regulation of osteoclastogenesis. However, the role of IL-3 in osteoclastogenesis remains controversial; whereas early studies showed that IL-3 stimulates osteoclastogenesis, recent investigations demonstrated that IL-3 inhibits osteoclast formation. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service