Skip to Content
MilliporeSigma

SML1184

OF-1

≥98% (HPLC)

Synonym(s):

4-Bromo-N-(2,3-dihydro-6-methoxy-1,3-dimethyl-2-oxo-1H-benzimidazol-5-yl)-2-methyl-benzenesulfonamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H18BrN3O4S
CAS Number:
Molecular Weight:
440.31
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352202
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

BrC1=CC(C)=C(S(NC(C(OC)=C2)=CC(N3C)=C2N(C)C3=O)(=O)=O)C=C1

InChI

1S/C17H18BrN3O4S/c1-10-7-11(18)5-6-16(10)26(23,24)19-12-8-13-14(9-15(12)25-4)21(3)17(22)20(13)2/h5-9,19H,1-4H3

InChI key

YUNQZQREIHWDQT-UHFFFAOYSA-N

Biochem/physiol Actions

OF-1 is a chemical probe for the bromodomains of the BRPF (BRomodomain and PHD Finger containing) family of scaffolding proteins (BRPF1, BRPF2, BRPF3) that assemble histone acetyltransferase (HAT) complexes of the MYST family members MOZ and MORF. The BRPF1 protein is the scaffold subunit of the MYST acetyltransferase complex, which plays a crucial roles in DNA repair, recombination and replication as well as transcription activation. OF-1 binds to BRPF1B with a Kd of 100 nM , to BRPF2 with a Kd of 500 nM and to BRPF3 with a Kd of 2.4 mM, and shows >100-fold selectivity against most other bromodomains. The closest off-target effects are against BRD4 (39-fold selectivity) and 50% inhibition of TIF1a at 20 μM. For full characterization details, please see OF-1 on the Structural Genomics Consortium (SGC) website.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc
OF-1 is a new SGC chemical probe for the bromodomains of the BRPF (BRomodomain and PHD Finger containing) family of scaffolding proteins (BRPF1/2/3).

Features and Benefits

OF-1 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.


Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Codes

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library