Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP3255

Sigma-Aldrich

TPO from rat

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

MGDF, Megakaryocyte colony-stimulating factor, Thrombopoietin, c-MPL Ligand

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.75

biological source

rat

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

≤0.2 ng/mL ED50

mol wt

18.7 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... THPO(81811)

General description

Thrombopoietin (TPO) also known as c-Mpl ligand is an acidic glycoprotein expressed mainly in the liver, kidney, and bone marrow. Recombinant rat TPO is a fully biologically active 174 amino acid polypeptide (18.7kDa), which contains the erythropoietin-like domain of the full length TPO protein.

Application

Thrombopoietin (TPO) from rat has been used in experimental systems to determine the effect of TPO in recovery from thrombocytopenia.

Biochem/physiol Actions

Thrombopoietin (TPO) acts as a key physiologic regulator of megakaryocyte and platelet production in vivo. Mutation of TPO or its receptor might lead to thrombocytopenia and megakaryocytopoiesis. In addition, TPO also influence the action of erythroid and erythromegakaryocytic progenitors in mouse embryonic yolk sac. TPO binds to c-mpl receptor and regulates circulating platelet level.

Sequence

SPVPPACDPR LLNKLLRDSY LLHSRLSQCP DVNPLSIPVL LPAVDFSLGE WKTQTEQSKA QDILGAVSLL LEGVMAARGQ LEPSCLSSLL GQLSGQVRLL LGALQGLLGT QLPPQGRTTA HKDPSALFLS LQQLLRGKVR FLLLVEGPAL CVRRTLPTTA VPSRTSQLLT LNKF

Physical form

Lyophilized from 5 mM Sodium Phosphate, pH 7.6 + 75 mM NaCl.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recombinant human thrombopoietin: basic biology and evaluation of clinical studies.
Kuter DJ and Begley CG
Blood, 100(10), 3457-3469 (2002)
Native thrombopoietin: structure and function.
Kato T
Stem Cells, 16(5), 322-328 (1998)
Thrombopoietin: the primary regulator of platelet production.
Kaushansky K
Blood, 86(2), 419-431 (1995)
Human thrombopoietin: gene structure, cDNA sequence, expression, and chromosomal localization.
Foster DC
Proceedings of the National Academy of Sciences of the USA, 91(26), 13023-13027 (1994)
Cloning and expression of murine thrombopoietin cDNA and stimulation of platelet production in vivo.
Lok S
Nature, 369(6481), 565-568 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service