Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3645

Sigma-Aldrich

LJH685

≥98% (HPLC)

Synonym(s):

2,6-Difluoro-4-(4-(4-(4-methylpiperazin-1-yl)phenyl)pyridin-3-yl)phenol, 2,6-Difluoro-4-[4-[4-(4-methyl-1-piperazinyl)phenyl]-3-pyridinyl]phenol, LJH 685, LJH-685, NVP-LJH685

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H21F2N3O
CAS Number:
Molecular Weight:
381.42
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

Fc1c(c(cc(c1)c2cnccc2c3ccc(cc3)N4CCN(CC4)C)F)O

InChI

1S/C22H21F2N3O/c1-26-8-10-27(11-9-26)17-4-2-15(3-5-17)18-6-7-25-14-19(18)16-12-20(23)22(28)21(24)13-16/h2-7,12-14,28H,8-11H2,1H3

Biochem/physiol Actions

LJH685 is potent pan-RSK inhibitor that targets RSK N-terminal kinase domain (NTKD) ATP-binding site (IC50/[ATP] = 6 nM/5 μM/RSK1, 5 nM/20 μM/RSK2, 4 nM/10 μM/RSK3). LJI308 effectivelfy inhibits MAPK pathway-dependent cancer growth (EC50 values of 0.73 μM/MDA-MB-231 and 0.79 μM/H358) by reducing the phosphorylation level of cellular Y-box binding protein-1 (YB1) Ser102 (EC50 = 0.2-0.3 μM; MDA-MB-231 and H358), but not that of ribosomal S6 protein (S6RP) S235/236.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaolin Guan et al.
Cell death discovery, 8(1), 84-84 (2022-02-27)
Breast cancer is one of the leading causes of mortality among women. Triple-negative breast cancer (TNBC) is responsible for a large percentage of all breast cancer deaths in women. This study demonstrated the function of Myb-like, SWIRM, and MPN domains
Dewei Jiang et al.
Cell death and differentiation, 29(6), 1283-1295 (2022-01-14)
Y-box binding protein 1 (YB-1) is a well-known oncogene highly expressed in various cancers, including basal-like breast cancer (BLBC). Beyond its role as a transcription factor, YB-1 is newly defined as an epigenetic regulator involving RNA 5-methylcytosine. However, its specific
Ida Aronchik et al.
Molecular cancer research : MCR, 12(5), 803-812 (2014-02-21)
The p90 ribosomal S6 kinase (RSK) family of serine/threonine kinases is expressed in a variety of cancers and its substrate phosphorylation has been implicated in direct regulation of cell survival, proliferation, and cell polarity. This study characterizes and presents the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service