Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP3056

Sigma-Aldrich

HVEM-Fc human

recombinant, expressed in Hi-5 Insect cells, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

ATAR, TNFSFR14, TR2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in Hi-5 Insect cells

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

1.3-1.9 μg/mL

mol wt

44.3 kDa

packaging

pkg of 100 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TNFRSF14(8764)

General description

HVEM belongs to the TNF Receptor superfamily of transmembrane proteins and plays a role in the activation of T-cells and other lymphocytes. It is expressed in various cells and tissues including spleen, thymus, lung, macrophages, and T-cells. HVEM activation induces a signaling cascade which results in induction of transcription factors NF-kappaB and AP-1. LIGHT (TNFSF14) and TNF-beta (TNFSF1) function as the ligands for HVEM, which can also bind specifically to herpes simplex virus glycoprotein D. Soluble HVEM can act as a "receptor decoy" resulting in inhibition of the activity of the HVEM ligands, LIGHT and TNF-beta. Recombinant human HVEM-Fc Chimera is a 376 amino acid fusion protein containing an N-terminal domain corresponding to the extracellular region of HVEM and a C-terminal domain corresponding to residues 102 to 330 of human IgG1.

Biochem/physiol Actions

HVEM belongs to the TNF Receptor superfamily of transmembrane proteins and plays a role in the activation of T-cells and other lymphocytes. Recombinant human HVEM-Fc Chimera is a 376 amino acid fusion protein containing an N-terminal domain corresponding to the extracellular region of HVEM and a C-terminal domain corresponding to residues 102 to 330 of human IgG1.

Sequence

LPSCKEDEYP VGSECCPKCS PGYRVKEACG ELTGTVCEPC PPGTYIAHLN GLSKCLQCQM CDPAMGLRAS RNCSRTENAV CGCSPGHFCI VQDGDHCAAC RAYATSSPGQ RVQKGGTESQ DTLCQNCPPG TFSPNGTLEE CQHQTKRSCD KTHTCPPCPA PELLGGPSVF LFPPKPKDTL MISRTPEVTC VVVDVSHEDP EVKFNWYVDG VEVHNAKTKP REEQYNSTYR VVSVLTVLHQ DWLNGKEYKC KVSNKALPAP IEKTISKAKG QPREPQVYTL PPSRDELTKN QVSLTCLVKG FYPSDIAVEW ESNGQPENNY KTTPPVLDSD GSFFLYSKLT VDKSRWQQGN VFSCSVMHEA LHNHYTQKSL SLSPGK

Physical form

Lyophilized from 1x PBS, pH 7.2.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1- 1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helene Baribault et al.
Expert opinion on therapeutic targets, 18(11), 1253-1264 (2014-10-08)
Although the human genome encodes ∼ 20,000 protein-coding genes, only a very small fraction of these have been explored as potential targets for therapeutic development. The challenge of identifying and validating new protein targets has contributed to the significant reduction
B S Kwon et al.
The Journal of biological chemistry, 272(22), 14272-14276 (1997-05-30)
The tumor necrosis factor receptor (TNFR) superfamily consists of approximately 10 characterized members of human proteins. We have identified a new member of the TNFR superfamily, TR2, from a search of an expressed sequence tag data base. cDNA cloning and
R I Montgomery et al.
Cell, 87(3), 427-436 (1996-11-01)
We identified and cloned a cellular mediator of herpes simplex virus (HSV) entry. Hamster and swine cells resistant to viral entry became susceptible upon expression of a human cDNA encoding this protein, designated HVEM (for herpesvirus entry mediator). HVEM was
L Yang et al.
Cell death & disease, 5, e1211-e1211 (2014-05-09)
ErbB2, an important membrane-bound receptor tyrosine kinase, was discovered nearly 30 years ago, but a natural ligand has never been found previously. ErbB2 is also an important oncogene and anticancer target, and its overexpression in cancer is associated with poor
Kenichiro Ito et al.
Nature communications, 6, 6373-6373 (2015-03-12)
Hepatocyte growth factor (HGF) receptor, also known as Met, is a member of the receptor tyrosine kinase family. The Met-HGF interaction regulates various signalling pathways involving downstream kinases, such as Akt and Erk. Met activation is implicated in wound healing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service