Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5127

Sigma-Aldrich

CD270 human

recombinant, expressed in E. coli, 0.5 mg protein/mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.75

biological source

human

recombinant

expressed in E. coli

description

0.1 mg recombinant human CD270 in 20 mM Tris-HCl buffer, containing NaCl, KCl, EDTA, L-arginine, DTT and glycerol.

sterility

Filtered sterilized solution

Assay

≥90% (SDS-PAGE)

form

liquid

packaging

pkg of 100 μg

concentration

0.5 mg protein/mL

accession no.

NP_003811.2

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... TNFRSF14(8764)

Application

Coating a plate well (6 well plate) with this recombinant CD270 protein in a specific culture medium at 5-10 μg/well allows for use 1) as a coating matrix protein for human T or B cell functions and differentiation regulation studies in vitro, 2) as potential biomarker protein for infectious diseases and auto-immuno disease diagnostic development, or 3) as an antigen for specific antibody production.

Use this procedure as a guideline to determine optimal coating conditions for the culture system of choice.
1. Thaw CD270 and dilute to desired concentration using serum-free medium or PBS. The final solution should be sufficiently dilute so the volume added covers the surface evenly (1-10 μg/well, 6 well plate).
2. Add appropriate amount of diluted material to culture surface.
3. Incubate at room temperature for approximately 1.5 hours.
4. Aspirate remaining material.
5. Rinse plates carefully with water and avoid scratching bottom surface of plates.
6. Plates are ready for use. They may also be stored at 2-8 °C damp or air dried if sterility is maintained.

Sequence

MASMTGGQQMGRGHHHHHHGNLYFQG^GEFLPSCKEDEYPVGSECCPKCSPGYRVKEACGELTGTVCEPCPPGTYIAHLNGLSKCLQCQMCDPAMGLRASRNCSRTENAVCGCSPGHFCIVQDGDHCAACRAYATSSPGQRVQKGGTESQDTLCQNCPPGTFSPNGTLEECQHQTKCSWLVTKAGAGTSSSHWV

Preparation Note

The extracellular domain of recombinant human CD270 (39 - 202 aa) was constructed with codon optimization and expressed with a small T7-His-TEV cleavage site Tag (29aa) fusion at its N-terminal and expressed in E. coli as inclusion bodies. The final product was refolded using our unique “temperature shift inclusion body refolding” technology and chromatographically purified.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Corinne Schaer et al.
PloS one, 6(4), e18495-e18495 (2011-05-03)
Tumor necrosis factor super family (TNFSF) members regulate important processes involved in cell proliferation, survival and differentiation and are therefore crucial for the balance between homeostasis and inflammatory responses. Several members of the TNFSF are closely associated with inflammatory bowel
B S Kwon et al.
The Journal of biological chemistry, 272(22), 14272-14276 (1997-05-30)
The tumor necrosis factor receptor (TNFR) superfamily consists of approximately 10 characterized members of human proteins. We have identified a new member of the TNFR superfamily, TR2, from a search of an expressed sequence tag data base. cDNA cloning and
R I Montgomery et al.
Cell, 87(3), 427-436 (1996-11-01)
We identified and cloned a cellular mediator of herpes simplex virus (HSV) entry. Hamster and swine cells resistant to viral entry became susceptible upon expression of a human cDNA encoding this protein, designated HVEM (for herpesvirus entry mediator). HVEM was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service