Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP0410

Sigma-Aldrich

ALOX15 active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥60% (SDS-PAGE)

Synonym(s):

15LOX-1, arachidonate 15-lipoxygenase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥60% (SDS-PAGE)

form

aqueous solution

mol wt

75 kDa

packaging

pkg of 20 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ALOX15(246)

General description

The gene ALOX15 (arachidonate 15-lipoxygenase) is mapped to human chromosome 17p13. It is mainly present in the epithelial cells in the upper airways, reticulocytes, eosinophils and macrophages. ALOX15 belongs to the dioxygenases family. It is an IL-4 (interleukin 4)/IL-13 target gene.
ALOX-15, also known as 15LOX-1, ω-6 lipoxygenase, or arachidonate 15-lipoxygenase (GenBank Accession No. NM_001140) amino acids 2- 662 (end) with N-terminal FLAG-tag, MW = 75kDa, expressed in Sf9 insect cells via a Baculovirus expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Biochem/physiol Actions

ALOX15 (arachidonate 15-lipoxygenase) is involved in homeostatic as well as pathological responses. ALOX15 is responsible for the oxidation of unsaturated fatty acids (at the 15 position) to active hydroperoxy and epoxy metabolites. It is also responsible for the conversion of linoleic acid to anti-tumor 13-S-hydroxyoctadecadienoic acid. ALOX15 is required for IL-4 (interleukin-4)/IL-13-mediated macrophage polarization. In mice, it is a negative regulator of bone mineral density. It is also involved in fatty acid metabolism. It controls MAPK (mitogen activated protein kinase) signaling in human airway epithelial cells using phosphatidylethanolamine-binding protein.

Physical form

Formulated in Tris-Buffered Saline (TBS), pH 7.4, containing 1 mM Fe(NH4)2(SO4)2.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymorphisms in inflammation associated genes ALOX15 and IL-6 are associated with bone properties in young women and fracture in elderly.
Herlin M, et al.
Bone, 79, 105-105 (2015)
12/15 Lipoxygenase regulation of colorectal tumorigenesis is determined by the relative tumor levels of its metabolite 12-HETE and 13-HODE in animal models.
Chang J, et al.
Oncotarget, 6, 2879-2879 (2015)
Interaction of human 15-lipoxygenase-1 with phosphatidylinositol bisphosphates results in increased enzyme activity.
Andersson E, et al.
Biochimica et Biophysica Acta, 1761, 1498-1498 (2006)
Wen G Jiang et al.
Prostaglandins, leukotrienes, and essential fatty acids, 74(4), 235-245 (2006-03-25)
15-lipoxygenase (15-LOX) belongs to the structurally and functionally related nonheme iron dioxygenases family. It has two isoforms, type-1 (leukocyte type) and type-2 (epidermis type) and converts arachidonic acid to eicosanoids including the anti-cancer 13-HODE. In the current study, we investigate
E Sigal et al.
Biochemical and biophysical research communications, 157(2), 457-464 (1988-12-15)
A full-length cDNA encoding 15-lipoxygenase has been isolated from a human reticulocyte cDNA library. The predicted primary structure of the enzyme exhibits a sequence similarity of 61% and 45% with human 5-lipoxygenase and the soybean lipoxygenase isoenzyme I, respectively. When

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service