Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4501034

Sigma-Aldrich

Anti-Arachidonate 5 Lipoxygenase antibody produced in rabbit

affinity isolated antibody

Synonym(s):

5-lipoxygenase, ALOX5, Arachidonate 5-lipoxygenase, LOG5, LOX5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 77 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ALOX5(240)

General description

The gene ALOX5 (arachidonate 5-lipoxygenase), also referred to as 5-LOX (5-lipoxygenase), encodes a protein belonging to the lipoxygenase family. The gene is mapped to human chromosome 10q11.21. The protein contains a distinct ‘C2-like′ domain at the amino terminal that is involved in Ca2+-dependent membrane binding and a large catalytic domain containing the non-heme catalytic iron. It is highly unstable with short-lived activity.

Immunogen

The antiserum was produced against synthesized peptide derived from human Arachidonate 5 Lipoxygenase.

Immunogen Range: 246-295

Biochem/physiol Actions

The gene ALOX5 (arachidonate 5-lipoxygenase) encodes a lipoxygenase that catalyzes the conversion of arachidonic acid (AA) and other fatty acids to bioactive compounds such as 5(S)-hydroxyeicosatetraenoic acid (5-HETE), leukotriene B4 (LTB4), and other leukotrienes. These leukotrienes and lipoxins function as mediators of inflammatory response. ALOX5 has been implicated in tumorigenesis and may serve as a potential therapeutic target and biomarker.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ALOX5 polymorphism associates with increased leukotriene production and reduced lung function and asthma control in children with poorly controlled asthma.
Mougey E
Clinical and Experimental Allergy, 43, 512-520 (2013)
The structure of human 5-lipoxygenase.
Gilbert NC
Science, 331, 217-219 (2011)
Arachidonate 5 lipoxygenase expression in papillary thyroid carcinoma promotes invasion via MMP-9 induction.
Kummer NT
Journal of Cellular Biochemistry, 113, 1998-2008 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service