Skip to Content
MilliporeSigma
All Photos(2)

Documents

SML3715

Sigma-Aldrich

SSK1

≥98% (HPLC)

Synonym(s):

(2R,3S,4S,5R,6S)-2-(acetoxymethyl)-6-(4-((((1-((2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-2-oxo-1,2-dihydropyrimidin-4-yl)carbamoyl)oxy)methyl)-2-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triyl triacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H34F2N4O18
CAS Number:
Molecular Weight:
788.61
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

SSK1 is a senescence cells specific killing agent that is activated by lysosomal β-galactosidase. SSK1 induces apoptosis in senescence cells through activation of p38 MAPK. Also, it induces mitochondrial DNA damage in senescent cells. In aged mice, SSK1 removes senescence cells in different tissues, attenuated low-grade inflammation and improved physical function.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Results of calf cartilage grafted in the human body].
S Y Wang
Zhonghua kou qiang yi xue za zhi = Zhonghua kouqiang yixue zazhi = Chinese journal of stomatology, 23(5), 282-283 (1988-09-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service