Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3621

Sigma-Aldrich

Erlotinib

≥98% (HPLC)

Synonym(s):

4-[(3-Ethynylphenyl)amino]-6,7-bis(2-methoxyethoxy)quinazoline, 6,7-Bis(2-methoxy-ethoxy)-quinazolin-4-yl]- (3-ethynyl-phenyl)amine, Erlotinib free base, N-(3-Ethynylphenyl)-6,7-bis(2-methoxyethoxy)-4-quinazolinamine, NSC 718781, OSI-774

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23N3O4
CAS Number:
Molecular Weight:
393.44
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

N(c3cc(ccc3)C#C)c1ncnc2c1cc(c(c2)OCCOC)OCCOC

InChI key

AAKJLRGGTJKAMG-UHFFFAOYSA-N

Biochem/physiol Actions

Erlotinib is an orally available, potent, and specific inhibitor of epidermal growth factor receptor (EGFR) tyrosine kinase. Erlotinib binds to the ATP binding site of the receptor. It is used to treat lung and pancreatic cancer.
orally available, potent, and specific inhibitor of epidermal growth factor receptor (EGFR) tyrosine kinase

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer Stamos et al.
The Journal of biological chemistry, 277(48), 46265-46272 (2002-08-28)
The crystal structure of the kinase domain from the epidermal growth factor receptor (EGFRK) including forty amino acids from the carboxyl-terminal tail has been determined to 2.6-A resolution, both with and without an EGFRK-specific inhibitor currently in Phase III clinical
Xiaoping He et al.
Scientific reports, 12(1), 8007-8007 (2022-05-15)
Hepatocellular carcinoma (HCC) is a leading cause of cancer-related death worldwide. Lenvatinib is approved as a first-line treatment for unresectable HCC. The therapeutic duration of lenvatinib is limited by resistance, but the underlying mechanism is unclear. To establish lenvatinib-resistant cells
Chike Osude et al.
Cells, 11(10) (2022-05-29)
NSCLC treatment includes targeting of EGFR with tyrosine kinase inhibitors (TKIs) such as Erlotinib; however, resistance to TKIs is commonly acquired through T790M EGFR mutations or overexpression of vascular endothelial growth factor receptor-2 (VEGFR-2). We investigated the mechanisms of EGFR-TKI

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service