Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3109

Sigma-Aldrich

Afatinib

≥98% (HPLC)

Synonym(s):

(2E)-N-[4-[(3-Chloro-4-fluorophenyl)amino]­-7-[[(3S)-tetrahydro-3-furanyl]oxy]-6-quinazolinyl]-4-(dimethylamino)-2-butenamide, (S,E)-N-(4-(3-Chloro-4-fluorophenylamino)-7-(tetrahydrofuran-3-yloxy)quinazolin-6-yl)-4-(dimethylamino)but-2-enamide, BIBW 2992, BIBW-2992, BIBW2992

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H25ClFN5O3
CAS Number:
Molecular Weight:
485.94
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN(C)C/C=C/C(NC1=CC(C(NC2=CC=C(C(Cl)=C2)F)=NC=N3)=C3C=C1O[C@H]4CCOC4)=O

InChI

1S/C24H25ClFN5O3/c1-31(2)8-3-4-23(32)30-21-11-17-20(12-22(21)34-16-7-9-33-13-16)27-14-28-24(17)29-15-5-6-19(26)18(25)10-15/h3-6,10-12,14,16H,7-9,13H2,1-2H3,(H,30,32)(H,27,28,29)/b4-3+/t16-/m0/s1

InChI key

ULXXDDBFHOBEHA-CWDCEQMOSA-N

Related Categories

Biochem/physiol Actions

Afatinib (BIBW2992) is an orally active, irreversible, potent and selective EGFR/HER2 (ErbB2) dual inhibitor (EGFR IC50 = 0.5 nM (Wt), 0.4 nM (L858R), 10 nM (L858R/T790M); HER2 IC50 = 14 nM) that covalently targets EGFR Cys773 and HER2 Cys805 residues. BIBW2992 suppresses 100 ng/mL EGF-induced EGFR/HER2 tyrosin phosphorylation (IC50 = 13-71 nM; A431, NIH-3T3-HER2, BT-474, NCI-N87), inhibits cancer cells survival in cultures and induces tumor regression in xenograft and transgenic lung cancer models in vivo (20 mg/kg/day p.o.).
Orally active, irreversible, potent and selective EGFR/HER2 (ErbB2) dual inhibitor with in vitro and in vivo anti-cancer efficacy.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT RE 2

Target Organs

Kidney,Skin

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service