Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3618

Sigma-Aldrich

KI-696

≥98% (HPLC)

Synonym(s):

(βS)​-β-​[3-​[[(4R)​-​3,​4-​Dihydro-​4-​methyl-​1,​1-​dioxido-​2H-​5,​1,​2-​benzoxathiazepin-​2-​yl]​methyl]​-​4-​methylphenyl]​-​7-​methoxy-​1-​methyl-1H-​benzotriazole-​5-​propanoic acid, (3S)-3-(7-Methoxy-1-methyl-1H-benzo[d][1,2,3]triazol-5-yl)-3-(4-methyl-3-(((R)-4-methyl-1,1-dioxido-3,4-dihydro-2H-benzo[b]-[1,4,5]oxathiazepin-2-yl)methyl)phenyl)propanoic acid, KI 696, KI696

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H30N4O6S
CAS Number:
Molecular Weight:
550.63
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[S]1(=O)(=O)N(C[C@H](Oc5c1cccc5)C)Cc2c(ccc(c2)[C@H](CC(=O)O)c3cc4nn[n](c4c(c3)OC)C)C

InChI key

ZDNGJXBUEQNFBQ-GCJKJVERSA-N

Biochem/physiol Actions

KI-696 is a non-cytotoxic (10 mM; BEAS-2B cells), Kelch domain-targeing (Kd ~1.3 nM) KEAP1 selective inhibitor that effectively upregulates nuclear NRF2 level in NHBE cells (%increase/[KI-696] = 100%/10 nM and 127%/100 nM; 6 hr) and NRF2 target genes transcription by blocking KEAP1-mediated NRF2 downregulation. KI-696 (5-50 mmol/kg i.v.) upregulates NRF2 target genes transcription in rat lungs in vivo, as well as alleviate ozone-induced pulmonary inflammation and GSH depletion. KI-696 inhibits OATP1B1, BSEP, and PDE3A only at much higher concentrations (IC50 = 2.5, 4.0, 10 mM, respectively) and exhibits little or no potency toward a panel of 46 channels, enzymes, and receptors.
Non-cytotoxic, selective KEAP1 Kelch domain inhibitor that effectively upregulates NRF2 actfivity in cultures and in rats in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Treatment of acute thromboembolism in hereditary antithrombin deficiency--a review].
L Tengborn
Lakartidningen, 84(15), 1249-1251 (1987-04-08)
Thomas G Davies et al.
Journal of medicinal chemistry, 59(8), 3991-4006 (2016-04-01)
KEAP1 is the key regulator of the NRF2-mediated cytoprotective response, and increasingly recognized as a target for diseases involving oxidative stress. Pharmacological intervention has focused on molecules that decrease NRF2-ubiquitination through covalent modification of KEAP1 cysteine residues, but such electrophilic
Patrizia Romani et al.
Nature cell biology, 24(2), 168-180 (2022-02-16)
Metastatic breast cancer cells disseminate to organs with a soft microenvironment. Whether and how the mechanical properties of the local tissue influence their response to treatment remains unclear. Here we found that a soft extracellular matrix empowers redox homeostasis. Cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service