Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3350

Sigma-Aldrich

Acriflavine purified

≥95% (HPLC)

Synonym(s):

Mixture of trypaflavines (3,6-diamino-10-methylacridinium chloride and 3,6-diamino-3-methylacridinehydrochloride) and proflavine (3,6-diaminoacridine) hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C13H11N3.HCl]x·[C14H13N3.HCl]y ·[C14H14N3·Cl]z
UNSPSC Code:
12352200
NACRES:
NA.25

Quality Level

Assay

≥95% (HPLC)

form

powder

color

red to brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC1=CC2=C(C=C1)C=C3C=CC(N)=CC3=N2.NC4=CC5=C(C=C4)C=C6C=CC(NC)=CC6=N5.NC7=CC8=C(C=C7)C=C9C=CC(N)=CC9=[N+]8C.[Cl-].Cl.Cl

InChI key

CNKPVFNEZPNSQB-UHFFFAOYSA-N

Biochem/physiol Actions

Acriflavine is an antimicrobial drug used to treat against sleeping sickness, urinary tract infections, and gonorrhea. Acriflavine is a potent inhibitor of SARS-CoV-2 papain-5 like protease (PLpro), which potently inhibits viral replication in vivo in mice and ex vivo in human airway epithelia. It is a mixture of trypaflavines (3,6-diamino-10-methylacridinium chloride and 3,6-diamino-3-methylacridinium chloride) and proflavine (3,6-diaminoacridine). Apparently commercial acriflavine mixtures contain the combination of differently methylated proflavines to optimally block the PLpro active site. Also, it potently inhibits replication of other betacoronaviruses.
Antimicrobial; potent inhibitor of SARS-CoV-2 papain-5 like protease (PLpro) and inhibits virus replication

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valeria Napolitano et al.
Cell chemical biology, 29(5), 774-784 (2022-01-13)
The COVID-19 pandemic caused by SARS-CoV-2 has been socially and economically devastating. Despite an unprecedented research effort and available vaccines, effective therapeutics are still missing to limit severe disease and mortality. Using high-throughput screening, we identify acriflavine (ACF) as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service