Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

239820

Sigma-Aldrich

AMD3100 octahydrochloride hydrate

≥98% (HPLC), solid, CXCR4 antagonist, Calbiochem

Synonym(s):

CXCR4 Antagonist I, AMD3100, 1,1ʹ-(1,4-Phenylene bis(methylene))- bis-1,4,8,11-tetraazacyclotetradecane, 8HCl, JM3100, 1,1ʹ-(1,4-Phenylenebis(methylene))-bis-1,4,8,11-tetraazacyclotetradecane, 8HCl, JM3100

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H54N8 · 8HCl
CAS Number:
Molecular Weight:
794.47
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Product Name

CXCR4 Antagonist I, AMD3100, The CXCR4 Antagonist I, AMD3100, also referenced under CAS 155148-31-5, controls the biological activity of CXCR4. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

off-white

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Cl.Cl.Cl.Cl.Cl.Cl.Cl.Cl.N3(CCNCCCNCCNCCC3)Cc1ccc(cc1)CN2CCNCCCNCCNCCC2

InChI

1S/C28H54N8.8ClH/c1-9-29-15-17-31-13-3-21-35(23-19-33-11-1)25-27-5-7-28(8-6-27)26-36-22-4-14-32-18-16-30-10-2-12-34-20-24-36;;;;;;;;/h5-8,29-34H,1-4,9-26H2;8*1H

InChI key

UEUPDYPUTTUXLJ-UHFFFAOYSA-N

General description

A symmetrical bicyclam compound that antagonizes CXCL12 (SDF1) binding to CXCR4 (IC50 = 108 and 245 nM using rat and human CXCR4, respectively) and inhibits SDF1-induced Ca2+ flux (by 100% at 100 ng/ml in SUP-T1 and THP-1 cultures) as well as CXCR4-, but not CCR5-, mediated HIV infection (IC50 ≤200 nM), while enhancing the binding of SDF1 to CXCR7 (by ~60% at 1 mM with CXCR7-expressing HEK293T cells) and SDF1-induced β-arrestin recruitment to CXCR7 (EC50 = 6.48 and 11.8 nM, in the presence and absence of 10 µM AMD3100, respectively). Administration AMD3100 via intravenous infusion is also reported to result in hematopoietic stem cell mobilization in humans, dogs, and mice in vivo. Exhibits no inhibitory effects against chemokine-induced signaling via CXCR1/2/3 or CCR1/2/3/4/5/6/7/8/9. Also available as a 50 mM solution in H2O (Cat. No. 239825).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Kalatskaya, I., et al. 2009. Mol. Pharmacol.77, 1240.
Pitchford, S.C., et al. 2009. Cell Stem Cell4, 62.
De Clercq, E. 2009. Biochem. Pharmacol. , 1655.
Thoma, G., et al. 2008. J. Med. Chem.51, 7915.
De Clercq, E. 2000. Mol. Pharmacol.57, 833.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sabrina Spiller et al.
Pharmaceutics, 13(10) (2021-10-24)
Biomaterials are designed to improve impaired healing of injured tissue. To accomplish better cell integration, we suggest to coat biomaterial surfaces with bio-functional proteins. Here, a mussel-derived surface-binding peptide is used and coupled to CXCL12 (stromal cell-derived factor 1α), a
Mark Soave et al.
Cell chemical biology, 27(10), 1250-1261 (2020-07-02)
Camelid single-domain antibody fragments (nanobodies) offer the specificity of an antibody in a single 15-kDa immunoglobulin domain. Their small size allows for easy genetic manipulation of the nanobody sequence to incorporate protein tags, facilitating their use as biochemical probes. The

Protocols

Learn how to perform cell migration assays in vitro using Millicell® hanging cell culture inserts and the suspension T-cell lines Jurkat and primary CD4+ cells. Monitor migration by flow cytometry and EZ-MTT assays.

Learn how to perform cell migration assays in vitro using Millicell® hanging cell culture inserts and the suspension T-cell lines Jurkat and primary CD4+ cells. Monitor migration by flow cytometry and EZ-MTT assays.

Learn how to perform cell migration assays in vitro using Millicell® hanging cell culture inserts and the suspension T-cell lines Jurkat and primary CD4+ cells. Monitor migration by flow cytometry and EZ-MTT assays.

Learn how to perform cell migration assays in vitro using Millicell® hanging cell culture inserts and the suspension T-cell lines Jurkat and primary CD4+ cells. Monitor migration by flow cytometry and EZ-MTT assays.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service